Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRGM Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116606 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-606 Supplier Abnova Corporation Supplier No. PAB13358
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IRGM.

Members of the p47 immunity-related GTPase (IRG) family, including IRGM, play an important role in the murine immune system; however, in humans this resistance system is greatly reduced. There is evidence that human IRGM plays a role in autophagy and control of intracellular mycobacteria (Bekpen et al., 2005; Singh et al., 2006 [PubMed 16888103]).[supplied by OMIM

Specifications

Antigen IRGM
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IRGM.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene IRGM
Gene Alias IFI1/IRGM1/LRG-47/LRG47/MGC149263/MGC149264
Gene Symbols IRGM
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region 14 amino acids of human IRGM.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345611
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.