Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA6 Rabbit anti-Horse, Human, Polyclonal Antibody, Abnova™

Catalog No. 89129282 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-282 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-282 Supplier Abnova Corporation Supplier No. PAB27764
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ITGA6.

The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen ITGA6
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ITGA6.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene ITGA6
Gene Alias CD49f/DKFZp686J01244/FLJ18737/ITGA6B/VLA-6
Gene Symbols ITGA6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acid at internal region of human ITGA6.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3655
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Equine, Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.