Learn More
ITGB4 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ITGB4.
Supplier: Abnova Corporation PAB18404
Description
Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
ITGB4 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
ITGB4 | |
ITGB4 | |
A synthetic peptide corresponding to residues surrounding Y1510 of human ITGB4. | |
100 μg | |
Primary | |
This antibody is specific to ITGB4. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of ITGB4. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
CD104 | |
Rabbit | |
Affinity purification | |
RUO | |
3691 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only