Learn More
ITPKA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ITPKA.
Supplier: Abnova Corporation PAB4051
Description
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro. ITPKA and ITPKB are 68% identical in the C-terminus region. [provided by RefSeq
Specifications
ITPKA | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ITPKA. | |
In PBS (0.09% sodium azide) | |
IP3KA | |
Rabbit | |
Protein G purification | |
RUO | |
3706 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
ITPKA | |
ITPKA | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ITPKA. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only