Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD1A (KDM3A) Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Kdm3a.
Supplier: Abnova Corporation PAB15824
Specifications
JMJD1A (KDM3A) | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Kdm3a. | |
In PBS (50% glycerol, 0.02% sodium azide) | |
1700105C21Rik/C230043E16Rik/JHDM2a/Jmjd1/KDM2A/TGSA/Tsga/mKIAA0742 | |
Rabbit | |
50 μg | |
Primary | |
Specific to recombinant protein GX0988. This antibody detects mJMJD1A protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Jmjd1a | |
Jmjd1a | |
Recombinant GST fusion protein corresponding to 167 amino acids of mouse Kdm3a. | |
RUO | |
104263 | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction