Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2D Rabbit anti-Human, Primates, Polyclonal Antibody, Abnova™

Catalog No. 89115783 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-783 Supplier Abnova Corporation Supplier No. PAB12409
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of JMJD2D.

Specifications

Antigen JMJD2D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of JMJD2D.
Dilution Western Blot (0.2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene JMJD2D
Gene Alias FLJ10251/KDM4D/MGC141909
Gene Symbols JMJD2D
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 460-520 of human JMJD2D.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55693
Target Species Human, Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.