Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kbtbd11, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117790 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-790 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-790 Supplier Abnova Corporation Supplier No. PAB15725
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Kbtbd11.

Sequence: LKYDPRRDEWQECPCSSSRERSADMVALDGFLYRFDLCGSRGEAQAAVGSGGGVSVFRYHCLAKQWSQCAVHLRPPGAPAGLQPFRCVALDGTIYCVSRAGTWRFVPSQDTEAGSDMGPGGSFEPEPLGSPLDVRGVLFPFVLNLPEKPDRGEQGAV

Specifications

Antigen Kbtbd11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Kbtbd11.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Kbtbd11
Gene Alias 4930465M17Rik/mKIAA0711
Gene Symbols Kbtbd11
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 157 mouse Kbtbd11.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74901
Test Specificity Specific to recombinant protein GX0626. This antibody detects mKBTBD11 protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.