Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCND2 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-632 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-632 Supplier Abnova Corporation Supplier No. PAB10106
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Kcnd2.

Specifications

Antigen KCND2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Kcnd2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.08% sodium azide)
Gene Kcnd2
Gene Alias Kv4.2/RK5/Shal1
Gene Symbols Kcnd2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH ) corresponding to amino acids 23-33 of Kcnd2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65180
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.