Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNIP1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118691 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-691 Supplier Abnova Corporation Supplier No. PAB16878
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Kcnip1.

Sequence: QTKQRRPSKDK

Specifications

Antigen KCNIP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Kcnip1.
Dilution Enzyme Immunoassay (1:300000) Western Blot (1:4000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene Kcnip1
Gene Alias 2900046L02Rik/3202002F18Rik/AI842357/AW494471/KCHIP1/Kchip1.2
Gene Symbols Kcnip1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to mouse Kcnip1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70357
Target Species Mouse
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.