Learn More
KCNIP3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KCNIP3.
Supplier: Abnova Corporation PAB18383
Description
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins, which belong to the recoverin branch of the EF-hand superfamily. Members of this family are small calcium binding proteins containing EF-hand-like domains. They are integral subunit components of native Kv4 channel complexes that may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. The encoded protein also functions as a calcium-regulated transcriptional repressor, and interacts with presenilins. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq
Specifications
KCNIP3 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
KCNIP3 | |
KCNIP3 | |
A synthetic peptide corresponding to residues surrounding S63 of human KCNIP3. | |
100 μg | |
Primary | |
This antibody is specific to KCNIP3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of KCNIP3. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
CSEN/DREAM/KCHIP3/MGC18289 | |
Rabbit | |
Affinity purification | |
RUO | |
30818 | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only