Learn More
KCNK13, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KCNK13.
Supplier: Abnova Corporation PAB25686
Description
This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming domains. The product of this gene is an open channel that can be stimulated by arachidonic acid. [provided by RefSeq
Specifications
KCNK13 | |
Polyclonal | |
Unconjugated | |
Western Blot (0.5 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
KCNK13 | |
KCNK13 | |
A synthetic peptide corresponding to 15 amino acids at internal region of human KCNK13. | |
100 μg | |
Primary | |
KCNK13 antibody is predicted to not cross-react with other KCNK protein family members. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of KCNK13. | |
In PBS (0.05% sodium azide) | |
K2p13.1/THIK-1/THIK1 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
56659 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only