Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK2, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117459 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-459 Supplier Abnova Corporation Supplier No. PAB15126
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.

This gene encodes one of the members of the two-pore-domain background potassium channel protein family. This type of potassium channel is formed by two homodimers that create a channel that leaks potassium out of the cell to control resting membrane potential. The channel can be opened, however, by certain anesthetics, membrane stretching, intracellular acidosis, and heat. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen KCNK2
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.
Dilution Western Blot (2 ug/ mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene KCNK2
Gene Alias K2p2.1/MGC126742/MGC126744/TPKC1/TREK/TREK-1/TREK1/hTREK-1c/hTREK-1e
Gene Symbols KCNK2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human KCNK2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3776
Test Specificity 88% sequence identity with rat protein. TREK 1 Please note that it is essential to run against membrane preps. NOT recommended for immunohistochemistry.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.