Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-704 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-704 Supplier Abnova Corporation Supplier No. PAB16891
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.

This gene encodes one of the members of the two-pore-domain background potassium channel protein family. This type of potassium channel is formed by two homodimers that create a channel that leaks potassium out of the cell to control resting membrane potential. The channel can be opened, however, by certain anesthetics, membrane stretching, intracellular acidosis, and heat. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: SASRERPGYTA

Specifications

Antigen KCNK2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.
Dilution EIA (1:500000) Western Blot (1:3000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene KCNK2
Gene Alias K2p2.1/MGC126742/MGC126744/TPKC1/TREK/TREK-1/TREK1/hTREK-1c/hTREK-1e
Gene Symbols KCNK2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human KCNK2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3776
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.