Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDR Rabbit anti-Human, DyLight 488, Polyclonal Antibody, Abnova™

Catalog No. 89117368 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-368 Supplier Abnova Corporation Supplier No. PAB14948
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Kdr.

Specifications

Antigen KDR
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Description Rabbit polyclonal antibody raised against synthetic peptide of Kdr.
Dilution Western Blot (1:250-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene Kdr
Gene Alias 6130401C07/Flk-1/Flk1/Krd-1/Ly73/VEGFR-2/VEGFR2
Gene Symbols Kdr
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1200-1300 of mouse Kdr.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16542
Test Specificity This antibody reacts with the human protein. Other species have not yet been tested. The immunogen used for this antibody production has 100% homology with mouse, rat and human proteins. This is specific to VEGR-2. This antibody is useful for Western blot analysis, where a doublet is seen at ∼150 KDa. Optimal working dilutions should be determined by the investigator.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.