Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137656 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-656 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-656 Supplier Abnova Corporation Supplier No. PAB13720
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length native Keratin.

Specifications

Antigen Keratin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length native Keratin.
Dilution Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In purified antiserum
Host Species Rabbit
Immunogen Native purified human Keratin.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes keratin.
Target Species Avian, Human, Mammalia
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.