Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Ki67 Antigen, Polyclonal, Abnova™

Catalog No. 89138354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-354 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-354 Supplier Abnova Corporation Supplier No. PAB18929

Specifications

Antigen Ki67 Antigen
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized from PBS (1% BSA, 0.09% sodium azide)
Host Species Rabbit
Immunogen Recombinant protein corresponding to a 1086 bp Ki67 motif-containing cDNA fragment of Ki67 Antigen.
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.