Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1377 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124979 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-979 Supplier Abnova Corporation Supplier No. PAB23281

Rabbit polyclonal antibody raised against recombinant KIAA1377.

Sequence: HKKMKYNIHERNGVRFLKSILKKESKYEHGYLKALIINQSFKFGNQKAAAIRDSIELTKEKGAEIPKTIKKLRWFDETSNIENNAENSHSLKNKTGTTQQHS

Specifications

Antigen KIAA1377
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description KIAA1377
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene KIAA1377
Gene Symbols KIAA1377
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human KIAA1377.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57562
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.