Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIDINS220 (phospho S919), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114725 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-725 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-725 Supplier Abnova Corporation Supplier No. PAB1025

Rabbit polyclonal antibody raised against synthetic phosphopeptide of KIDINS220.

Specifications

Antigen KIDINS220
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description kinase D-interacting substrate, 220kDa
Formulation In PBS (0.02% sodium azide)
Gene KIDINS220
Gene Alias ARMS, MGC163482
Gene Symbols KIDINS220
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S919 of human KIDINS220.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57498
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.