Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF11 Rabbit anti-Human, Mammals, Mouse, Pig, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115782 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-782 Supplier Abnova Corporation Supplier No. PAB12408
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant KIF11.

This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis. [provided by RefSeq

Specifications

Antigen KIF11
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant KIF11.
Dilution Immunofluorescence (1:1000) Immunoprecipitation (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene KIF11
Gene Alias EG5/HKSP/KNSL1/TRIP5
Gene Symbols KIF11
Host Species Rabbit
Immunogen Recombinant protein corresponding to coiled-coil region of human KIF11.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3832
Test Specificity This antibody is specific to Eg5.
Target Species Human, Mammalia, Mouse, Pig, Primate, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.