Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF1A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121798 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-798 Supplier Abnova Corporation Supplier No. PAB20294

Rabbit polyclonal antibody raised against recombinant KIF1A.

The protein encoded by this gene is a member of the kinesin family. This protein is highly similar to mouse heavy chain kinesin member 1A protein which is an anterograde motor protein that transports membranous organelles along axonal microtubules. It is thought that this protein may play a critical role in the development of axonal neuropathies resulting from impaired axonal transport. There are multiple polyadenylation sites found in this gene. [provided by RefSeq

Sequence: SGTAKISFDDQHFEKFQSESCPVVGMSRSGTSQEELRIVEGQGQGADVGPSADEVNNNTCSAVPPEGLLLDSSEKAALDGPLDAALDHLRLGNTFTFRVTVLQASSISAEYADIF

Specifications

Antigen KIF1A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant KIF1A.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene KIF1A
Gene Alias ATSV/C2orf20/DKFZp686I2094/FLJ30229/HUNC-104/MGC133285/MGC133286/UNC104
Gene Symbols KIF1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human KIF1A.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 547
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.