Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF5C Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Kif5c.
Supplier: Abnova Corporation PAB15721
Description
Sequence: VKALESALKEAKENAMRDRKRYQQEVDRIKEAVRAKNMARRAHSAQIAKPIRPGHYPASSPTAVHAVRGGGGGSSNSTHYQKSpecifications
KIF5C | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Kif5c. | |
In PBS (50% glycerol, 0.02% sodium azide) | |
KINN/Khc/NKHC/NKHC-2/NKHC2/mKIAA0531 | |
Rabbit | |
50 μg | |
Primary | |
Specific to recombinant protein GX0067. This antibody detects endogenous mKIF5C protein in several cell types. It also recognizes human KIF5C protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Kif5c | |
Kif5c | |
Recombinant GST fusion protein corresponding to 82 mouse Kif5c. | |
RUO | |
16574 | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction