Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIT, Rabbit, Clone: YR145, Abnova™

Catalog No. 89108967 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-967 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-967 Supplier Abnova Corporation Supplier No. MAB10766
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against KIT.

This gene encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen KIT
Applications Immunohistochemistry
Classification Monoclonal
Clone YR145
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against KIT.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein base, 0.09% sodium azide)
Gene KIT
Gene Alias C-Kit/CD117/PBT/SCFR
Gene Symbols KIT
Host Species Rabbit
Immunogen KIT.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3815
Content And Storage Store at 4°C is stable for 3 years.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.