Learn More
KIT (phospho Y719), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of KIT.
Supplier: Abnova Corporation PAB16884
Description
This gene encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: SDSTNEYpMDMKPGSpecifications
KIT | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of KIT. | |
Lyophilized from 10mM PBS | |
C-Kit/CD117/PBT/SCFR | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Lyophilized |
ELISA | |
Unconjugated | |
ELISA (1:64000) The optimal working dilution should be determined by the end user. | |
KIT | |
KIT | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y719 of KIT. | |
RUO | |
3815 | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only