Learn More
KITLG Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes the ligand of the tyrosine-kinase receptor encoded by the KIT locus. This ligand is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | KITLG |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of KITLG. |
| Dilution | Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS (0.02% sodium azide) |
| Gene | KITLG |
| Gene Alias | DKFZp686F2250/KL-1/Kitl/MGF/SCF/SF/SHEP7 |
| Gene Symbols | KITLG |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.