Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK12 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-681 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-681 Supplier Abnova Corporation Supplier No. PAB1018

Rabbit polyclonal antibody raised against synthetic peptide of KLK12.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Alternate splicing of this gene results in three transcript variants encoding different isoforms. [provided by RefSeq

Sequence: KYVDWIRMIMRNN

Specifications

Antigen KLK12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description kallikrein-related peptidase 12
Formulation In PBS (0.02% sodium azide)
Gene KLK12
Gene Alias DKFZp686H1078, KLK-L5, KLKL5, MGC42603
Gene Symbols KLK12
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 236-248 of human KLK12.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43849
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.