Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLK14 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a mixture of synthetic peptide conjugated to KLH.
Supplier: Abnova Corporation PAB0137
Specifications
KLK14 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a mixture of synthetic peptide conjugated to KLH. | |
Lyophilized from PBS, pH 7.4 | |
A mixture of synthetic peptides (conjugated to KLH) corresponding to KLK14. | |
RUO | |
Human | |
Lyophilized |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (6 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at -20°C on dry atmosphere. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction