Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK4 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114668 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-668 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-668 Supplier Abnova Corporation Supplier No. PAB1015

Rabbit polyclonal antibody raised against synthetic peptide of KLK4.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. In some tissues its expression is hormonally regulated. The expression pattern of a similar mouse protein in murine developing teeth supports a role for the protein in the degradation of enamel proteins. Alternate splice variants for this gene have been described, but their biological validity has not been determined. [provided by RefSeq

Sequence: KFTEWIEKTVQAS

Specifications

Antigen KLK4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description kallikrein-related peptidase 4
Formulation In PBS (0.02% sodium azide)
Gene KLK4
Gene Alias ARM1, EMSP, EMSP1, KLK-L1, MGC116827, MGC116828, PROSTASE, PRSS17, PSTS
Gene Symbols KLK4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 242-254 of human KLK4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9622
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.