Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK9 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114677 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-677 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-677 Supplier Abnova Corporation Supplier No. PAB1017

Rabbit polyclonal antibody raised against synthetic peptide of KLK9.

KLK9 belongs to the kallikrein subgroup of serine proteases, which have diverse physiologic functions in many tissues. For background information on kallikreins, see MIM 147960.[supplied by OMIM

Sequence: HYLDWIQEIMEN

Specifications

Antigen KLK9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description kallikrein-related peptidase 9
Formulation In PBS (0.02% sodium azide)
Gene KLK9
Gene Alias KLK-L3, KLK8, KLKL3
Gene Symbols KLK9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 239-250 of human KLK9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284366
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.