Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-KLRG1, Alexa Fluor 488, Clone: 1151A, R&D Systems™

Catalog No. FAB6944G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6944G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6944G025 Supplier R&D Systems Supplier No. FAB6944G025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Antigen KLRG1
Applications Flow Cytometry
Classification Monoclonal
Clone 1151A
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O88713
Gene Alias 2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
Gene Symbols KLRG1
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived mouse KLRG1, Glu57-Tyr188, Accession # O88713
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10219
Test Specificity Detects mouse KLRG1 in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.