Learn More
KPNA4 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KPNA4.
Supplier: Abnova Corporation PAB3804
Description
The nuclear import of karyophilic proteins is directed by short amino acid sequences termed nuclear localization signals (NLSs). Karyopherins, or importins, are cytoplasmic proteins that recognize NLSs and dock NLS-containing proteins to the nuclear pore complex. The protein encoded by this gene shares the sequence similarity with Xenopus importin-alpha and Saccharomyces cerevisiae Srp1. This protein is found to interact with the NLSs of DNA helicase Q1 and SV40 T antigen. [provided by RefSeq
Specifications
KPNA4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of KPNA4. | |
In PBS (0.09% sodium azide) | |
IPOA3/MGC12217/MGC26703/QIP1/SRP3 | |
Rabbit | |
Protein G purification | |
RUO | |
3840 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
KPNA4 | |
KPNA4 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human KPNA4. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only