Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KREMEN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89130773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-773 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-773 Supplier Abnova Corporation Supplier No. PAB2991
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of KREMEN1.

This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq

Specifications

Antigen KREMEN1
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of KREMEN1.
Dilution Western Blot (1:1000) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene KREMEN1
Gene Alias FLJ31863/KREMEM1/KREMEN/KRM1
Gene Symbols KREMEN1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human KREMEN1.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83999
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.