Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT16, Rabbit, Clone: R20-S, Abnova™

Catalog No. 89113768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-768 Supplier Abnova Corporation Supplier No. MAB9767
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of KRT16.

The protein encoded by this gene is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region of chromosome 17q12-q21. This keratin has been coexpressed with keratin 14 in a number of epithelial tissues, including esophagus, tongue, and hair follicles. Mutations in this gene are associated with type 1 pachyonychia congenita, nonepidermolytic palmoplantar keratoderma and unilateral palmoplantar verrucous nevus. [provided by RefSeq

Specifications

Antigen KRT16
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone R20-S
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of KRT16.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene KRT16
Gene Accession No. P08779
Gene Alias CK16/K16/K1CP/KRT16A/NEPPK
Gene Symbols KRT16
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human KRT16.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3868
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.