Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT17, Rabbit, Clone: SP95, Abnova™

Catalog No. 89109245 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-245 Supplier Abnova Corporation Supplier No. MAB11031

Rabbit monoclonal antibody raised against synthetic peptide of KRT17.

This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex. [provided by RefSeq

Specifications

Antigen KRT17
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SP95
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of KRT17.
Dilution Western Blot (1:25) Immunohistochemistry (1:100) Flow Cytometry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene KRT17
Gene Alias K17/PC/PC2/PCHC1
Gene Symbols KRT17
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human KRT17.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3872
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.