Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT18, Rabbit, Clone: R20-H, Abnova™

Catalog No. 89113770 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-770 Supplier Abnova Corporation Supplier No. MAB9769
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of KRT18.

KRT18 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen KRT18
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone R20-H
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of KRT18.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:300) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene KRT18
Gene Accession No. P05783
Gene Alias CYK18/K18
Gene Symbols KRT18
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human KRT18.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3875
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.