Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT5, Rabbit, Clone: SP27, Abnova™

Catalog No. 89110846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-846 Supplier Abnova Corporation Supplier No. MAB2146

Rabbit monoclonal antibody raised against synthetic peptide of KRT5.

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. [provided by RefSeq

Specifications

Antigen KRT5
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SP27
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of KRT5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Flow Cytometry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide).
Gene KRT5
Gene Alias CK5/DDD/EBS2/K5/KRT5A
Gene Symbols KRT5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human KRT5.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3852
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.