Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT7, Rabbit, Clone: R17-S, Abnova™

Catalog No. 89110553 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-553 Supplier Abnova Corporation Supplier No. MAB1657
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of KRT7.

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the simple epithelia lining the cavities of the internal organs and in the gland ducts and blood vessels. The genes encoding the type II cytokeratins are clustered in a region of chromosome 12q12-q13. Alternative splicing may result in several transcript variants; however, not all variants have been fully described. [provided by RefSeq

Specifications

Antigen KRT7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone R17-S
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of KRT7.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene KRT7
Gene Alias CK7/K2C7/K7/MGC129731/MGC3625/SCL
Gene Symbols KRT7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human KRT7.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.