Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KTN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89122641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-641 Supplier Abnova Corporation Supplier No. PAB21149
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant KTN1.

Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin (see MIM 148760 and MIM 600025), an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.[supplied by OMIM

Sequence: ELKRLEAMLKERESDLSSKTQLLQDVQDENKLFKSQIEQLKQQNYQQASSFPPHEELLKVISEREKEISGLWNELDSLKDAVEHQRKKNNERQQQVEAVELEAKE

Specifications

Antigen KTN1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant KTN1.
Dilution Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene KTN1
Gene Alias CG1/KIAA0004/KNT/MGC133337/MU-RMS-40.19
Gene Symbols KTN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human KTN1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3895
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.