Learn More
L3MBTL Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against L3MBTL.
Supplier: Abnova Corporation PAB14105
Description
This gene encodes the homolog of a protein identified in Drosophila as a suppressor of malignant transformation of neuroblasts and ganglion-mother cells in the optic centers of the brain. This gene product is localized to condensed chromosomes in mitotic cells. Overexpression of this gene in a glioma cell line results in improper nuclear segregation and cytokinesis producing multinucleated cells. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
L3MBTL | |
Polyclonal | |
Rabbit polyclonal antibody raised against L3MBTL. | |
In PBS (0.05% sodium azide, 0.05% proclin 300) | |
DKFZp586P1522/FLJ41181/H-L(3)MBT/KIAA0681/L3MBTL1/dJ138B7.3 | |
Rabbit | |
50 μg | |
Primary | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
L3MBTL | |
L3MBTL | |
N-terminus of human L3MBTL. | |
RUO | |
26013 | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only