Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lacI, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114730 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-730 Supplier Abnova Corporation Supplier No. PAB10255
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant lacI.

Specifications

Antigen lacI
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant lacI.
Dilution ELISA (1:90000-1:100000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene lacI
Gene Alias ECK0342/JW0336
Gene Symbols lacI
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length Escherichia coli (strain DH10B) lacI.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 945007
Test Specificity This antibody reacts with recombinant Lac I protein. A BLAST analysis was used to suggest cross-reactivity with Lac I from DNA-binding transcriptional repressor in Escherichia coli (strain DH10B) and transcriptional repressor of the lac operon in Shigella sonnei (strain Ss046) based on a 100% homology with the immunizing sequence.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.