Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LACTB Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89124688 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-688 Supplier Abnova Corporation Supplier No. PAB22989
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant LACTB.

This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins. [provided by RefSeq

Sequence: NSNENLLPGYLKPETMVMMWTPVPNTEMSWDKEGKYAMAWGVVEKKQTYGSCRKQRHYASHTGGAVGASSVLLVLPEELDTETINNKVP

Specifications

Antigen LACTB
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant LACTB.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene LACTB
Gene Alias FLJ14902/G24/MRPL56
Gene Symbols LACTB
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human LACTB.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114294
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.