Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lama1, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-400 Supplier Abnova Corporation Supplier No. PAB15005
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Lama1.

Specifications

Antigen Lama1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against native Lama1.
Dilution Western Blot (1:100-1:5000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:2000) Immunohistochemistry (Frozen sections) (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Lama1
Gene Alias AA408497/Lama
Gene Symbols Lama1
Host Species Rabbit
Immunogen Native purified Lama1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16772
Test Specificity human, mouse and rat laminin. Other species have not been tested. This is pan-specific and reacts well with all laminin isoforms tested: laminin-1 (alpha-1, beta-1, and gamma-1) and laminin-2 (alpha-2, beta-1, and gamma-1) . NB 300-144 can be used for Western blotting where it recognizes a band at 200 and 400 KDa, representing laminin.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.