Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lama1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-631 Supplier Abnova Corporation Supplier No. PAB12133
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Lama1.

Specifications

Antigen Lama1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Lama1.
Dilution Immunocytochemistry (1:50-1:200) Immunohistochemistry (Frozen sections) (1:500-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:2000) Western Blot (1:100-1:5000) The optimal working dilution should be deter
Formulation In PBS (0.05% sodium azide)
Gene Lama1
Gene Alias AA408497/Lama
Gene Symbols Lama1
Host Species Rabbit
Immunogen Native purified Lama1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16772
Test Specificity This antibody is pan-specific and reacts well with all laminin isoforms tested: laminin-1 (alpha-1, beta-1, and gamma-1) and laminin-2 (alpha-2, beta-1, and gamma-1).
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.