Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-LAMA4, Polyclonal, Abnova™

Catalog No. 89128563 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-563 Supplier Abnova Corporation Supplier No. PAB26919
Add to Cart
Add to Cart

Specifications

Antigen LAMA4
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of LAMA4.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene LAMA4
Gene Alias DKFZp686D23145/LAMA3/LAMA4*-1
Gene Symbols LAMA4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human LAMA4.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3910
Test Specificity LAMA4 polyclonal antibody detects endogenous levels of LAMA4 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.