Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89138352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-352 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-352 Supplier Abnova Corporation Supplier No. PAB18925

Rabbit polyclonal antibody raised against native Lambda Light Chain.

Specifications

Antigen Lambda Light Chain
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Lambda Light Chain.
Dilution Immunohistochemistry (1:1000-1:2000) Western Blot (1:3000-1:4000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Host Species Rabbit
Immunogen Native purified Lambda Light Chain from human sera.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.