Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116848 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-848 Supplier Abnova Corporation Supplier No. PAB13805
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of LAMP2.

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may play a role in tumor cell metastasis. It may also function in the protection, maintenance, and adhesion of the lysosome. Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins. [provided by RefSeq

Specifications

Antigen LAMP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of LAMP2.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene LAMP2
Gene Accession No. NP_034815
Gene Alias CD107b/LAMPB/LGP110
Gene Symbols LAMP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region 17 amino acids of human LAMP2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Test Specificity Recognizes LAMP-2.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.