Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARS Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89130031 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-031 Supplier Abnova Corporation Supplier No. PAB28521
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant LARS.

This gene encodes a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. The encoded enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain. Alternatively spliced transcript variants of this gene have been found; however, their full-length nature is not known. [provided by RefSeq

Specifications

Antigen LARS
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant LARS.
Dilution Western Blot (1:4000) Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene LARS
Gene Alias FLJ10595/FLJ21788/HSPC192/KIAA1352/LARS1/LEURS/LEUS/LRS/PIG44/RNTLS/hr025Cl
Gene Symbols LARS
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 930-1176 of human LARS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51520
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.