Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LASS2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114636 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-636 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-636 Supplier Abnova Corporation Supplier No. PAB10110
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of LASS2.

This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq

Specifications

Antigen LASS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of LASS2.
Dilution Western blot (1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene LASS2
Gene Alias CerS2/FLJ10243/L3/MGC987/SP260/TMSG1
Gene Symbols LASS2
Host Species Rabbit
Immunogen A synthetic peptide of human LASS2.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29956
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.