Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT2 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115954 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-954 Supplier Abnova Corporation Supplier No. PAB1262
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Lat2.

Specifications

Antigen LAT2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Lat2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Lat2
Gene Alias AW125574/LAB/NTAL/WSCR5/Wbscr15/Wbscr5
Gene Symbols Lat2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 124-203 of mouse Lat2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56743
Test Specificity This antibody recognizes N-terminal cytoplasmic domain of mouse Non-T cell Activation Linker (NTAL), also known as LAB (linker of activated B cells).
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.