Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LATS1/LATS2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LATS1/LATS2.
Supplier: Abnova Corporation PAB18515
Specifications
LATS1/LATS2 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
WARTS/wts | |
Rabbit | |
Affinity purification | |
RUO | |
9113/26524 | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of LATS1/LATS2. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
LATS1 | |
A synthetic peptide corresponding to human LATS1/LATS2. | |
100 μg | |
Primary | |
This antibody is specific to LATS1/LATS2. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction