Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Lcat, Polyclonal,

Catalog No. ABS1621MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1621MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1621MI Supplier MilliporeSigma™ Supplier No. ABS1621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Lcat in Human, Mouse samples, and it is validated for Western Blotting

Phosphatidylcholine-sterol acyltransferase (EC 2.3.1.43; UniProt P16301; also known as Lecithin-cholesterol acyltransferase, Phospholipid-cholesterol acyltransferase, LCAT) is encoded by the Lcat (also known as AI046659; D8Wsu61e) gene (Gene ID 16816) in murine species. LCAT catalyzes the esterification of cholesterol and phosphatidylcholines (PCs) to cholesteryl esters and lysoPCs on the surface of high and low density lipoproteins (HDLs and LDLs). LCAT reaction is essential for the maturation of nascent HDL particles in plasma, and a genetic deficiency of LCAT results in the near absence of normal HDL particles and the presence of plasma very low density and low density lipoprotein particles of abnormal size and shape. LCAT is initially produced with a 24-aa N-terminal signal peptide, which is later cleaved to form the mature enzyme. LCAT is synthesized in the liver before it is secreted into the plasma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lcat
Applications Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P16301
Gene Symbols Lcat
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Lcat.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_032516
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.